Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TSH (Human) ELISA Kit

Catalog No. p-4748197 Shop All Abnova Corporation Products
Click to view available options
Sample Type:
Serum
Calibration Range:
0 to 10uIU/mL
0 to 25uIU/mL
0.05 to 12μIU/mL
0.2 to 40μIU/mL

Includes

  • Murine Monoclonal Anti-TSH coated Microtiter wells, 1 plate, 96 wells
  • Reference Standard Set: Contains 0, 0.1, 0.5, 2, 5, and 10μIU/mL, lyophilized
  • Enzyme Conjugate Reagent, 13mL
  • TMB Reagent (One-Step), 11mL
  • Stop Solution (1N HCl), 11mL

The ELISA Test is based on principle of a solid phase enzyme-linked immunosorbent assay

Specifications

Calibration Range 0 to 10uIU/mL
Conjugate HRP
Cross Reactivity Human
Description TSH (Human) High Sensitive ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of thyroid stimulating hormone.
Detection Limit 0.07uIU/mL
Detection Method Colorimetric
Includes Murine Monoclonal Anti-TSH coated Microtiter wells. 1 plate, 96 wells, Reference Standard Set: Contains 0, 0.1, 0.5, 2, 5, and 10μIU/ml. Lyophilized, Enzyme Conjugate Reagent (13mL), TMB Reagent (One-Step) (11mL), Stop Solution (1N HCl) (11mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.