Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Visual hCG Pregnancy ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
Includes
- Microtiter Wells: mouse monoclonal anti-α-hCG coated wells, 96 wells
- Enzyme Conjugate: containing mouse monoclonal anti-β-hCG peroxidase conjugate in protein stabilizer, 7mL (Red cap)
- HCG Standard: containing 0mIU/mL hCG, 1mL (White cap)
- HCG Standard: containing 20mIU/mL hCG, 1mL (Yellow cap)
- HCG Standard: containing 150mIU/mL hCG, 1mL (Black cap)
- TMB Reagent (One-Step), 7mL (Amber cap)
- Stop Solution: 1N HCl, 7mL (Natural cap)
The method employs two monoclonal antibodies to selectively identify hCG in urine/serum with a high degree of sensitivity
Specifications
Specifications
Description | Visual hCG Pregnancy ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human chorionic gonadotropin. |
Quantity | 1 Kit |
Regulatory Status | For research use only (RUO) |
Sensitivity | 20 mIU/mL |
Packaging | 96-well |
Content And Storage | Store the kit at 4°C |
For Use With (Application) | Quantification |
Product Type | ELISA Kit |
No. of Assays | 96 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction