Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABO, Blood Group A Antigen Antibody (HE-14) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2478680.2MG
Description
ABO, Blood Group A Antigen Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Agglutination, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
ABO, Blood Group A Antigen | |
Monoclonal | |
1.0 mg/mL | |
Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, Agglutination | |
A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT | |
Mouse | |
Protein A or G purified | |
RUO | |
28 | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Immunofluorescence, Agglutination | |
HE-14 | |
Unconjugated | |
PBS with No Preservative | |
ABO | |
Mixture of erythrocytes of blood group A1 and glycoprotein fraction isolated from the saliva of secretors with blood group A | |
0.2 mg | |
Primary | |
This antibody recognizes human blood group A (monofucosyl and difucosyl A antigens with chain types 1, 2, 3, 4, 5, 6) and Forssmann antigen. It is also reactive with the immuno-dominant A trisaccharide. Blood group antigen expression in human colon cancer was studied by means of two monoclonal antibodies of broad anti-A (HE-14) and anti-type 3 and type 4 chain-based A and H (HE-10) specificity. These antigens were proved to re-appear in tumors of the distal colon, the HE-10 antibody reacting more frequently (9 out of 12 samples) than HE-14 (5 out of 12 samples) and frequently with supra-nuclear staining of the cytoplasm probably in those places of the Golgi apparatus where carbohydrate antigens are synthesized. This staining pattern is characteristic of HE-10 in normal colonic mucosa as well. With HE-14, staining was often absent in less differentiated tumors, while HE-10 did react in such tumors. In some cases, these two antibodies gave different staining patterns in parallel sections from the same tissue sample, primarily at the cellular level. Three out of 12 cases showed blood group antigen expression in the mucosa of the distal colon adjacent to the tumor only when HE-10 Ab was used. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction