Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABO, Blood Group A Antigen Antibody (HE-195), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311705
Description
ABO, Blood Group A Antigen Monoclonal specifically detects ABO, Blood Group A Antigen in Human samples. It is validated for Flow Cytometry.Specifications
| ABO, Blood Group A Antigen | |
| Monoclonal | |
| 1 mg/ml | |
| Flow Cytometry 1-4 ug/ml | |
| A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT | |
| Mixture of erythrocytes of group A1 and glycoprotein fraction isolated from saliva of secretors with blood group A. | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgM |
| Flow Cytometry | |
| HE-195 | |
| Unconjugated | |
| TBS | |
| Mouse | |
| Multi-step | |
| RUO | |
| 28 | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction