Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABO, Blood Group B Antigen Antibody (HEB-29), Novus Biologicals™
SDP

Catalog No. p-200007040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 ml
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1202524 1 mL
NB228781 0.25 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1202524 Supplier Novus Biologicals Supplier No. NB1202524
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

ABO, Blood Group B Antigen Monoclonal specifically detects ABO, Blood Group B Antigen in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Aggregation, Agglutination.

Specifications

Antigen ABO, Blood Group B Antigen
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Aggregation
Classification Monoclonal
Clone HEB-29
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Aggregation
Gene Accession No. P16442
Gene Alias A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT
Gene Symbols ABO
Host Species Mouse
Immunogen Mixture of erythrocytes of blood group B and glycoprotein fraction isolated from the saliva of secretors with blood group B.
Purification Method Concentrated Supernatant
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28
Test Specificity The antibody HEB-29 reacts with human blood group B. The specifity of the antibody HEB-29 was confirmed by comparison of specifity and reactivity to standard reagent using >5.000 samples of blood.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.