Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABO Recombinant Rabbit Monoclonal Antibody (2V4R0)
GREENER_CHOICE

Catalog No. PIMA535675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535675 Supplier Invitrogen™ Supplier No. MA535675
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MAEVLRTLAG KPKCHALRPM ILFLIMLVLV LFGYGVLSPR SLMPGSLERG FCMAVREPDH LQRVSLPRMV YPQPKVLTPC RKDVLVVTPW LAPIVWEGTF.

Histo-blood group ABO system transferase (NAGAT) is a member of the glycosyltransferase 6 family of proteins and the basis of the ABO blood group system. The histo-blood group ABO involves three carbohydrate antigens: A, B and H; the NAGAT protein functions as the basis of this group. A, B and AB individuals express a glycosyltransferase activity converting the H antigen to the A antigen (by addition of UDP-GalNAc) or to the B antigen (by addition of UDP-Gal), while O individuals do not express this glycosyltransferase activity. The B phenotype of NAGAT differs from the A form by a few residue substitutions, whereas the O form is a result of a single base frame-shift deletion in the N-terminal extremity of the gene. The NAGAT protein localizes to the Golgi apparatus and its conserved DXD motif functions in cofactor binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABO
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2V4R0
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ABO
Gene Accession No. P16442
Gene Alias a transferase; A1-specific alpha 1-3-N-acetylgalactosaminyltransferase; A3GALNT; A3GALT1; ABO; ABO A3 transferase; ABO blood group (alpha 1-3-N-acetylgalactosaminyltransferase, alpha 1-3-galactosyltransferase); ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase, transferase B, alpha 1-3-galactosyltransferase); ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase); ABO glycosyltransferase; ABO weak transfer; Abo2; B (A) alpha 1; B blood group galactosyltransferase; b transferase; B(A) alpha-1,3-galactosyltransferase; BGAT; blood group A glycosyltransferase 2; Cis-AB transferase; cis-AB transferase 2; fucosylglycoprotein; fucosylglycoprotein 3-alpha-galactosyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form; Glycoprotein-fucosylgalactoside alpha-galactosyltransferase; glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase; glycosyltransferase A; glycosyltransferase B; GTB; Histo-blood group A transferase; histo-blood group A2 transferase; histo-blood group ABO system transferase; Histo-blood group ABO system transferase 2; histo-blood group B transferase; NAGAT; NAGAT 2; putative blood group A transferase T2; Transferase; transferase A; transferase B, alpha 1-3-galactosyltransferase)
Gene Symbols ABO
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human BGAT (P16442).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.