Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABO Mouse anti-Human, Unconjugated, Clone: HE-14, Abnova™

Catalog No. 89337518 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-518 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-518 Supplier Abnova Corporation Supplier No. MAB21025
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ABO Monoclonal antibody specifically detects ABO in Human samples. It is validated for IHC-P, IF.

Specifications

Antigen ABO
Applications Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone HE-14
Conjugate Unconjugated
Description ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
Gene ABO
Gene Alias A3GALNT, A3GALT1, GTB, NAGAT
Host Species Mouse
Immunogen Mixture of erythrocytes of blood group A1 and glycoprotein fraction isolated from the saliva of secretors with blood group A
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 28
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.