Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563028 Supplier Invitrogen™ Supplier No. PA563028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EESEASPQVH PFPDHELEDM KMKISALKSE IQKEKANKGQ SRAIERLKKK MFENEFLLL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene encodes a protein that is similar to the protein encoded by the breakpoint cluster region gene located on chromosome 22. The protein encoded by this gene contains a GTPase-activating protein domain, a domain found in members of the Rho family of GTP-binding proteins. Functional studies in mice determined that this protein plays a role in vestibular morphogenesis, suggesting that Rho-related GTPases help coordinate motor skills and balance. Alternatively spliced transcript variants that encode different isoforms have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABR
Gene Accession No. Q12979
Gene Alias ABR; active BCR-related; active BCR-related gene; active breakpoint cluster region-related protein; MDB
Gene Symbols ABR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABR. Recombinant protein control fragment (Product #RP-101395).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.