Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abraxas Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176827MFV500
Description
Abraxas Polyclonal antibody specifically detects Abraxas in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Abraxas | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BRCA1-A complex subunit Abraxas, ABRA1, CCDC98, family with sequence similarity 175, member A | |
Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human CCDC98. The immunogen is located within amino acids 80 - 130 of CCDC98. Amino Acid Squence: WYKFRRHSDQIMTFR | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
84142 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction