Learn More
Invitrogen™ Abscisic acid Recombinant Mouse Monoclonal Antibody (4G7)
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA566653
Description
Abscisic acid Recombinant Monoclonal Antibody for ELISA
Specifications
| Abscisic acid | |
| Recombinant Monoclonal | |
| 6.4 mg/mL | |
| PBS with 0.09% sodium azide | |
| Conjugate of ABA hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C | |
| Liquid |
| ELISA | |
| 4G7 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.