Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Absolute Q™ dPCR SARS-CoV-2 Wastewater Surveillance Kit

Catalog No. A55241
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55241 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A55241 Supplier Applied Biosystems™ Supplier No. A55241
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems™ Absolute Q™ dPCR SARS-CoV-2 Wastewater Surveillance Kit enables fast, sensitive, and absolute quantification of SARS-CoV-2 from isolated wastewater nucleic acid using the QuantStudio™ Absolute Q™ Digital PCR System.

The Applied Biosystems Absolute Q dPCR SARS-CoV-2 Wastewater Surveillance Kit enables fast, sensitive, absolute quantification of SARS-CoV-2 from isolated wastewater nucleic acid using the QuantStudio Absolute Q Digital PCR System.

Features of the Absolute Q dPCR SARS-CoV-2 wastewater surveillance kit include:
• Ease of use—simple qPCR-like workflow with just a few pipetting steps and minimal hands-on time
• Fast—absolute quantification of targets of interest in less than two hours, including reaction preparation and dPCR data collection
• Consistency—powered by microfluidic array plate (MAP) technology and TaqMan chemistry for exceptional data consistency

The Absolute Q SARS-CoV-2 Wastewater Surveillance Kit contains both a triplex digital PCR (dPCR) assay and Absolute Q 1-Step RT-dPCR Master Mix (Cat. No. A55146). The high-resolution multiplex assay targets two SARS-CoV-2 N gene (nucleoplasmid protein) regions because the gene is the more stable and less susceptible to mutations. The assay was designed by our experts with experience in rapidly responding to the SARS-CoV-2 outbreak, who helped ensure SARS-CoV-2 specificity and followed CDC guidance.

In accordance with CDC guidelines for SARS-CoV-2 wastewater surveillance, an assay targeting pepper mild mottle virus (PMMoV) is also part of the single tube triplex assay to enable fecal normalization. The assay enables all three targets to be detected in a single well. With the QuantStudio Absolute Q dPCR System, the red channel is free to enable further multiplexing with your own target of interest by combining the assay kit with an appropriate dye-labeled assay.

The wastewater surveillance kit offers enough multiplex assay and master mix for 200 sample reactions and is for use with the QuantStudio Absolute Q MAP16 Plate.

Related products
QuantStudio Absolute Q dPCR System (Cat. No. A52864)
QuantStudio Absolute Q MAP16 Plate kit (Cat. No. A52865)
MagMAX Wastewater Ultra Nucleic Acid Isolation Kit (Cat. No. A52606)
MagMAX Wastewater Ultra Nucleic Acid Isolation Kit with Virus Enrichment (Cat. No. A52610)
AcroMetrix SARS-CoV-2 Control, RUO (Cat. No. 954517)
AcroMetrix Coronavirus 2019 (COVID-19) RNA Control, RUO (Cat. No. 954519)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Absolute Q 1-step RT-dPCR Master Mix (4X)
• Absolute Q Wastewater Surveillance Assay (20X)

Store at –15 to –25°C.

Format Tube
PCR Method Digital PCR
Reaction Speed <2 hr.
Technique Digital PCR
For Use With (Equipment) QuantStudio™ Absolute Q™ Digital PCR System
Label or Dye FAM, VIC, ABY
Quantity 200 reactions
Shipping Condition Dry Ice
Target SARS-CoV-2 Pepper Mild Mottle Virus (PMMoV)
For Use With (Application) Pathogen Detection
Hot Start Hot Start Taq Polymerase
No. of Reactions 200
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.