Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ABsolute QPCR Mix, ROX

Catalog No. AB1138B
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
1600 Reactions
400 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
AB1138B 1600 Reactions
AB-1139A 400 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB1138B Supplier Thermo Scientific™ Supplier No. AB1138B
Add to Cart
Add to Cart

Optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes.

Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes.

  • High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
  • Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
  • Optimized buffer - For maximal performance in multiplex reactions
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 16 x 1.25 mL ABsolute QPCR ROX Mix (2X)

Store at -20°C
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
Product Line ABsolute
Product Type qPCR Mix
Quantity 1,600 reactions
Sufficient For 1600 Reactions
For Use With (Application) Gene Expression
Fidelity (vs. Taq) 2 X
No. of Reactions 1600 Reactions
Sample Type DNA, dsDNA
Volume 20 mL
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.