Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ABsolute QPCR Mix, SYBR Green, low ROX

Catalog No. AB1323A Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
400 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
AB-1323-A 400 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB-1323-A Supplier Thermo Scientific™ Supplier No. AB1323A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized for SYBR Green chemistry and contains all the components necessary to perform quantitative PCR, with the exception of template and primers.

The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.

ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.

  • High Sensitivity & Specificity - The master mixes contain the patented hot start enzyme Thermo-Start DNA Polymerase
  • Optimized buffer containing SYBR Green
  • Reproducibility - All master mixes undergo extensive QC to ensure batch to batch consistency
  • Broad dynamic range
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 1 x 5 mL ABsolute QPCR SYBR Green Low ROX Mix (2X)
• 100 μL MgCl2 (1M)

Store at -20°C
Detection Method SYBR
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
Product Line ABsolute
Product Type Real Time PCR SYBR Master Mix
Quantity 400 reactions
Sufficient For 400 Reactions of 25 μL Each
For Use With (Application) Gene Expression
Compatibility qPCR Cyclers
No. of Reactions 400 Reactions
Sample Type DNA, dsDNA
Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.