Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ABT 089 Dihydrochloride
GREENER_CHOICESDP

Catalog No. 507950R Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
507950R 50 mg
507910R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 507950R Supplier R&D Systems™ Supplier No. 5079/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. High affinity and selective α4β2 nAChR partial agonist

ABT 089 dihydrochloride is a high affinity and selective α4β2 partial agonist (Ki = 16 nM). Exhibits >1000-fold and >10,000-fold selectivity for α4β2 over α1β1γ1 and α7 receptors respectively. Neuroprotective against glutamate-induced toxicity in rat cortical neurons in vitro. Enhances cognitive performance in vivo. Orally bioavailable and brain penetrant.

Specifications

Chemical Name or Material ABT 089 dihydrochloride
CAS 161416-61-1
Synonym 2-Methyl-3-[(2S)-pyrrolidinylmethoxy]pyridine dihydrochloride
Quantity 50 mg
Solubility Soluble to 100mM in water and to 20mM in DMSO
Product Line Tocris
Molecular Formula C11H16N2O·2HCl
Formula Weight 265.18
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.