Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222333
Description
ABT1 Polyclonal specifically detects ABT1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ABT1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
activator of basal transcription 1, Basal transcriptional activator, hABT1, TATA-binding protein-binding protein | |
Rabbit | |
Affinity Purified | |
RUO | |
29777 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
NP_037507.1 | |
ABT1 | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Activator of Basal Transcription 1 using the numbering given in entry NP_037507.1 (GeneID 29777). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction