Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585017
Description
Immunogen sequence: SKKRVVPGIV YLGHIPPRFR PLHVRNLLSA YGEVGRVFFQ AEDRFVRRKK KAAAAAGGKK RSYTKDYTEG WVEFR.
Could be a novel TATA-binding protein (TBP) which can function as a basal transcription activator. Can act as a regulator of basal transcription for class II genes.
Specifications
ABT1 | |
Polyclonal | |
Unconjugated | |
Abt1 | |
ABT1; Activator of basal transcription 1; basal transcriptional activator; hABT1; hypothetical protein LOC553568; im:7139827; TATA-binding protein-binding protein; zgc:109995 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29777 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULW3 | |
Abt1 | |
Recombinant protein corresponding to Human ABT1. Recombinant protein control fragment (Product #RP-108674). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction