Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112348
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112348 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112348 Supplier Invitrogen™ Supplier No. PA5112348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABT1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Could be a novel TATA-binding protein (TBP) which can function as a basal transcription activator. Can act as a regulator of basal transcription for class II genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABT1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Abt1
Gene Accession No. Q9QYL7, Q9ULW3
Gene Alias ABT1; Activator of basal transcription 1; basal transcriptional activator; hABT1; hypothetical protein LOC553568; im:7139827; TATA-binding protein-binding protein; zgc:109995
Gene Symbols Abt1
Host Species Rabbit
Immunogen Recombinant Human Activator of basal transcription 1 protein (197-270AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29777, 30946
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.