Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABTB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565075 Supplier Invitrogen™ Supplier No. PA565075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVDTRISVRIH EYAAISLTAC MENLVEEIRA RVMASHSPDG GGAGGGEVSA EALEMVINND AELWGVLQPY EH Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

May be involved in the initiation of hepatocyte growth. Target protein displays a transient increase in mRNA expression prior to DNA synthesis in regenerating liver cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABTB2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABTB2
Gene Accession No. Q8N961
Gene Alias ABTB2; ABTB2A; ankyrin repeat and BTB (POZ) domain containing 2; ankyrin repeat and BTB domain containing 2; ankyrin repeat and BTB/POZ domain-containing protein 2; BTBD22
Gene Symbols ABTB2
Host Species Rabbit
Immunogen Recombinant Human ABTB2. Recombinant protein control fragment (Product #RP-105837).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25841
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.