Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AC 186
GREENER_CHOICE

Catalog No. 505310RD
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-531-0RD 10 mg
50-535-0RD 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-531-0RD Supplier Tocris Bioscience™ Supplier No. 5053/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ERβ agonist; neuroprotective

AC 186 is a potent and selective ERβ agonist (EC50 values are 6 and 5000 nM for ERβ and ERα respectively). Exhibits gender selective neuroprotective effects in a male rat model of Parkinson's disease. Decreases Aβ levels in combination with ACP-105 in a rat model of Alzheimer's disease.

Specifications

Quantity 10 mg
Product Type AC 186
Purity >99%
Molecular Weight (g/mol) 306.32

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.