Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AC 1903
GREENER_CHOICESDP

Catalog No. 67665 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-665 5 mg
67-662-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-665 Supplier Tocris Bioscience™ Supplier No. 6766/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective TRPC5 blocker

AC 1903 is a selective TRPC5 blocker (IC50 = 13.6 μM). Also displays weak inhibition at TRPC4 (IC50 >100 μM) but not TRPC6. Exhibits no effect against a standard panel of profiling kinases. Blocks TRPC5 channel activity in glomeruli of proteinuric rats. Suppresses severe proteinuria and prevents podocyte loss in a transgenic rat model of focal segmental glomerulosclerosis (FSGS).

Specifications

Chemical Name or Material N-(2-Furanylmethyl)-1-(phenylmethyl)-1H-benzimidazol-2-amine
CAS 831234-13-0
Quantity 5 mg
Molecular Formula C19H17N3O
Formula Weight 303.36
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.