Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AC 261066
GREENER_CHOICESDP

Catalog No. 404650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-465-0 50 mg
40-461-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-465-0 Supplier R&D Systems™ Supplier No. 4046/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. RARβ2 agonist

Potent RARβ2 agonist (pEC50 = 8.1). Selective over RARβ1, RARα and RARγ (pEC50 values are 6.4, 6.2 and 6.3 respectively). More potent and orally available than AC 55649 (Cat. No. 2436).

Chemical Identifiers

CAS 870773-76-5
Molecular Formula C17H20FNO4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 353.41
Purity >98%
Molecular Weight (g/mol) 353.408
Chemical Name or Material AC 261066
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.