Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AC 264613
GREENER_CHOICESDP

Catalog No. 337050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-705-0 50 mg
33-701-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-705-0 Supplier R&D Systems™ Supplier No. 3370/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PAR2 receptor agonist

Potent and selective protease-activated receptor 2 (PAR2) agonist (pEC50 = 7.5). Displays no activity at other PAR subtypes and exhibits no significant activity at over 30 other receptors implicated in nociception and inflammation. Stimulates PI hydrolysis, Ca2+ mobilization and cellular proliferation in vitro (pEC50 values are 6.9, 7.0 and 7.5 respectively).

Chemical Identifiers

CAS 1051487-82-1
Molecular Formula C19H18BrN3O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 400.27
Purity >98%
Molecular Weight (g/mol) 400.276
Chemical Name or Material AC 264613
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.