Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AC 55541
SDP

Catalog No. 336950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-695-0 50 mg
33-691-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-695-0 Supplier R&D Systems™ Supplier No. 3369/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective PAR2 agonist

Potent and selective protease-activated receptor 2 (PAR2) agonist (pEC50 = 6.7) that displays no activity at other PAR subtypes or at over 30 other receptors involved in nociception and inflammation. Stimulates cell proliferation, PI hydrolysis and Ca2+ mobilization in vitro (pEC50 values are 6.7, 5.9 and 6.6 respectively) and exhibits pronociceptive activity in vivo.

Chemical Identifiers

CAS 916170-19-9
Molecular Formula C25H20BrN5O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 518.36
Purity >97%
Molecular Weight (g/mol) 518.371
Chemical Name or Material AC 55541
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.