Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AC 710
GREENER_CHOICE

Catalog No. 501310RD
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-131-0RD 10 mg
50-135-0RD 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-131-0RD Supplier Tocris Bioscience™ Supplier No. 5013/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PDGFR family inhibitor

AC 710 is a potent PDGFR family inhibitor (IC50 values are 1.2, 2, 7.7 and 10.5 nM for KIT, FLT3, PDGFRβ and CSF1R respectively; Kd = 1.3 nM for PDGFRα). Exhibits >30-fold selectivity over a panel of other kinases and exhibits no activity against cytochrome P450. Causes tumor regression of leukemia cell xenografts in mice. Also reduces joint swelling and inflammation in a mouse model of collagen-induced arthritis. Orally active.

Specifications

Quantity 10 mg
Product Type AC 710
Purity >99%
Molecular Weight (g/mol) 562.7

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.