Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ac-Cys-NHMe
GREENER_CHOICESDP

Catalog No. 793610 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-361-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 79-361-0 Supplier R&D Systems™ Supplier No. 7936/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Building block for the lysine acylation using conjugating enzymes (LACE) technique

Ac-Cys-NHMe is a non-canonical amino acid that is an essential building block for lysine acylation using conjugating enzymes (LACE) technique. LACE is a chemoenzymatic approach that uses an E2 small ubiquitin-like modifier (SUMO)-conjugating enzyme, Ubc9, to conjugate peptide thioesters containing a C-terminal ubiquitin-derived sequence (LRLRGG) to a lysine residue located in a recognition sequence (IKQE) on a protein via an isopeptide bond. Ac-Cys-NHMe can be reacted with a peptide hydrazide to form a reactive peptide thioester, which is the substrate for Ubc9. LACE enables the site-specific modification of proteins at lysine residues to allow attachment of biochemical probes such as fluorophores to proteins or to engineer proteins for site-specific delivery.

Specifications

Chemical Name or Material (2R)-2-(Acetylamino)-3-mercapto-N-methylpropanamide
CAS 10061-65-1
Quantity 10 mg
Product Line Tocris
Target Unnatural Amino Acids
Molecular Formula C6H12N2O2S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.