Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Checkers AC Series Aviation Wheel Chocks

Catalog No. 19088479 Shop All Justrite Safety Group Products
Click to view available options
Color:
Lime Green
Yellow
Includes:
Locking Rope
Loop Rope

Catalog No. 19-088-479

Supplier: Checkers AC6820QLRECHFLG

Only null left
Add to Cart
Add to Cart

Lightweight polyurethane aviation wheel chocks are ideal for private, commercial, cargo, and military aircraft.

  • Checkers™ Aviation Wheel Chocks are made of highly durable, lightweight polyurethane
  • AC Chocks last longer than wood chocks, never need painting and eliminate Foreign Object Damage threats for aircraft
  • The AC Series is ideal for private, commercial, cargo and military aircraft

Specifications

Dimensions (L x W x H) 20 x 8 x 6.5 in.
Material Urethane
For Use With (Equipment) Private, Commercial, Cargo, and Military Aircrafts
Includes Loop Rope
Color Lime Green
Product Type Wheel Chock
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.