Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ AC800 ESD Nitrile-Coated Cut Resistant Gloves

Catalog No. 18139740
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large

Electrostatic dissipative 18-gauge HPPE, cut resistant embossed microporous foam nitrile coating glove

The new AC800 is an 18-gauge seamless knit glove made using HPPE blended with static dissipative yarn and featuring an embossed microporous foam nitrile palm dip. The unique features of these elements allow for this product to be touchscreen friendly, silicone free, fingerprint free and contains no natural rubber latex. Certified under the standard EN 16350, the AC800 by SHOWA meets the requirements for electrostatic properties (ESD). This certification ensures the glove’s safe usage in ATEX zones (working area with risk of explosion and/or high flammability).

Specifications

Accelerator Free Yes
Applications Industrial
Coating Material Nitrile Foam
Color Gray
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material HPPE
Powdered Powder Free
Product Type ESD Gloves
Size Large
Sterility Non-sterile
Thickness (English) 18 gauge
Thickness (Metric) 0.8 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.