Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ac4GalN6yne
GREENER_CHOICE

Catalog No. 793210
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-321-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 79-321-0 Supplier Tocris Bioscience™ Supplier No. 7932/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Alkyne containing monosaccharide building block; used in BOCTAG

Ac4GalN6yne is a cell-permeable alkyne-containing monosaccharide building block. It can be used in Bio-Orthogonal Cell-specific Tagging of Glycoproteins (BOCTAG) technique. In this strategy, cells carrying an artificial biosynthetic pathway (B. longum NahK and mut-AGX1) can generate alkyne-tagged UDP-GalN6yne, that are then used by glycosyltransferases to chemically tag the glycoproteome. Ac4GalN6yne can be used for bioorthogonal labelling in vitro and mouse models.

Specifications

Chemical Name or Material 2-Deoxy-2-[(1-oxo-5-hexyn-1-yl)amino]-D-galactopyranose-1,3,4,6-tetraacetate
Quantity 10 mg
Target Glycobiology Probes
Molecular Formula C20H27NO10
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.