Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ac4ManNAz
GREENER_CHOICESDP

Catalog No. 747910 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-791-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 74-791-0 Supplier R&D Systems™ Supplier No. 7479/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Azide-containing monosaccharide building block; used as glycoprotein, glycolipid and glycoRNA probe

Ac4ManNAz is a cell-permeable, unnatural azide-containing monosaccharide building block. Ac4ManNAz is metabolized and incorporated as azidosialic acid into cell surface and secreted sialoglycoproteins to allow Cu(I)-free click chemistry for further conjugation or detection. Ac4ManNAz can be used for bioorthogonal labeling of human prostate tissue slice cultures; the target sialoglycoproteins can be biotinylated for proteomic analysis or reacted with a cyclooctyne-functionalized optical probe for imaging. Ac4ManNAz is also used to label glycoRNAs on the cell surface.

Specifications

Chemical Name or Material 2-[(2-Azidoacetyl)amino]-2-deoxy-D-mannopyranose-1,3,4,6-tetraacetate
CAS 361154-30-5
Quantity 10 mg
Product Line Tocris
Target Photoactivatable Cross-linking Probes
Molecular Formula C16H22N4O10
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.