Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ac4ManNDAz
GREENER_CHOICESDP

Catalog No. 755310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-531-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 75-531-0 Supplier Tocris Bioscience™ Supplier No. 7553/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Photo-crosslinking probe for studying glycoproteins and glycolipids

Ac4ManNDAz is a cell-permeable photo-crosslinking probe for studying glycoprotein or glycolipid interactions. The compound is an unnatural diazirine-containing monosaccharide building block, which acts as a photo-crosslinker. Once inside the cell, Ac4ManNDAz is metabolized and incorporated into cell surface glycoproteins as a photo-crosslinking sialic acid analog. Upon UV irradiation, the diazirine group is activated to a reactive carbene that covalently crosslinks to nearby molecules.

Specifications

Chemical Name or Material 2-Deoxy-2-[[3-(3-methyl-3 H-diazirin-3-yl)-1-oxopropyl]amino]-D-mannopyranose-1,3,4,6-tetraacetate
CAS 1015698-39-1
Quantity 10 mg
Target Photoactivatable Cross-linking Probes
Molecular Formula C19H27N3O10
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.