Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AC9 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | AC9 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-7792
![]() |
Bethyl Laboratories, Inc
A304465AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Adenylyl cyclases function to convert ATP to cyclic AMP in response to activation by a variety of hormones, neurotransmitters and other regulatory molecules. Cyclic AMP, in turn, activates several other target molecules (primarily cyclic AMP-dependent protein kinases) to control a broad range of diverse phenomena such as metabolism, gene transcription and memory. Classically, adenylyl cyclases respond to receptor-initiated signals, mediated by the Gs and Gi heterotrimeric G proteins. The binding of an agonist to a Gs-coupled receptor (i.e., alpha beta-adrenergic receptor) catalyzes the exchange of GDP (bound to G alpha s) for GTP, dissociation of GTP-G alpha sfrom G beta gamma and G alpha s-mediated activation of adenylyl cyclase. The most abundant cerebral adenylyl cyclases appears to be adenylyl cyclase IX. AC IX is confined to the gray matter and its expression is mainly neuronal, with its highest expression located at the hippocampus. ACIX is also expressed in heart, pancreas and thyrocytes. AC I and AC IX are regulated reciprocally by intracellular free Ca2+. The inhibition of AC IX by Ca2+ is blocked by the calcineurin inhibitors FK506 and cyclosporin A.Specifications
AC9 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
AC9 | |
Between 1303 and 1353 | |
Primary | |
4° C | |
ADCY9 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O60503, P51830 | |
115, 11515 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title