Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AC9 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501567790
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7790 Supplier Bethyl Laboratories, Inc Supplier No. A304465A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Adenylyl cyclases function to convert ATP to cyclic AMP in response to activation by a variety of hormones, neurotransmitters and other regulatory molecules. Cyclic AMP, in turn, activates several other target molecules (primarily cyclic AMP-dependent protein kinases) to control a broad range of diverse phenomena such as metabolism, gene transcription and memory. Classically, adenylyl cyclases respond to receptor-initiated signals, mediated by the Gs and Gi heterotrimeric G proteins. The binding of an agonist to a Gs-coupled receptor (i.e., alpha beta-adrenergic receptor) catalyzes the exchange of GDP (bound to G alpha s) for GTP, dissociation of GTP-G alpha sfrom G beta gamma and G alpha s-mediated activation of adenylyl cyclase. The most abundant cerebral adenylyl cyclases appears to be adenylyl cyclase IX. AC IX is confined to the gray matter and its expression is mainly neuronal, with its highest expression located at the hippocampus. ACIX is also expressed in heart, pancreas and thyrocytes. AC I and AC IX are regulated reciprocally by intracellular free Ca2+. The inhibition of AC IX by Ca2+ is blocked by the calcineurin inhibitors FK506 and cyclosporin A.

Specifications

Antigen AC9
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ADCY9
Gene Accession No. O60503, P51830
Gene Alias AC9
Gene Symbols Adcy9
Host Species Rabbit
Immunogen Between 1303 and 1353
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115, 11515
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.