Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAA1 Antibody (CL2663), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00
Specifications
Antigen | ACAA1 |
---|---|
Clone | CL2663 |
Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
Classification | Monoclonal |
Description
ACAA1 Monoclonal antibody specifically detects ACAA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
ACAA1 | |
Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P09110 | |
30 | |
IgG1 | |
Protein A purified |
CL2663 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
Mouse | |
Proteases & Other Enzymes, Signal Transduction | |
PBS (pH 7.2), 40% Glycerol | |
Acetyl-CoA acyltransferase, acetyl-CoA acyltransferase 1, EC 2.3.1, peroxisomal | |
This antibody was developed against Recombinant Protein corresponding to amino acids: VLKDVNLRPEQLGDICVGNVLQPGAGAIMARIAQFLSDIPETVPLSTVNRQCSSGLQAVASIAGGIRNGSYDIGMACGVESMSLADRGNPGNITSRLMEKEKARDCLIPMGITSENVAERFGISREKQ | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title