Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACAA1 Monoclonal Antibody (CL2662)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531372
Description
Immunogen sequence: VLKDVNLRPE QLGDICVGNV LQPGAGAIMA RIAQFLSDIP ETVPLSTVNR QCSSGLQAVA SIAGGIRNGS YDIGMACGVE SMSLADRGNP GNITSRLMEK EKARDCLIPM GITSENVAER FGISREKQ.
This gene encodes an enzyme operative in the beta-oxidation system of the peroxisomes. Deficiency of this enzyme leads to pseudo-Zellweger syndrome. Alternative splicing results in multiple transcript variants.
Specifications
ACAA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09110 | |
ACAA1 | |
Recombinant protein corresponding to Human ACAA1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
CL2662 | |
Unconjugated | |
ACAA1 | |
3-ketoacyl-CoA thiolase A, peroxisomal; 3-ketoacyl-CoA thiolase B, peroxisomal; 3-ketoacyl-CoA thiolase, peroxisomal; ACAA; ACAA1; Acaa1a; Acaa1b; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 1; acetyl-CoA acyltransferase 1A; acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A 1 peroxisomal; Acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A peroxisomal; acetyl-CoA acyltransferase A; Acetyl-CoA acyltransferase B; acetyl-CoA acyltransferase, 3-oxo acyl-CoA thiolase A; acetyl-Coenzyme A acyltransferase 1; acetyl-Coenzyme A acyltransferase 1 (peroxisomal 3-oxoacyl-Coenzyme A thiolase); acetyl-Coenzyme A acyltransferase 1A; acetyl-Coenzyme A acyltransferase 1B; beta-ketothiolase; Beta-ketothiolase A; beta-ketothiolase B; D9Ertd25e; peroxisomal 3-ketoacyl-CoA thiolase; peroxisomal 3-ketoacyl-CoA thiolase B; peroxisomal 3-oxoacyl-CoA thiolase; Peroxisomal 3-oxoacyl-CoA thiolase A; peroxisomal 3-oxoacyl-CoA thiolase B; peroxisomal 3-oxoacyl-Coenzyme A thiolase; Pktaa; PTHIO; PTL; RGD1562373; testicular tissue protein Li 197; THIO; thiolase A; thiolase B | |
Mouse | |
Protein A | |
RUO | |
30 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction