Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACAA1 Monoclonal Antibody (CL2663)
GREENER_CHOICE

Catalog No. PIMA534995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534995 Supplier Invitrogen™ Supplier No. MA534995
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: VLKDVNLRPE QLGDICVGNV LQPGAGAIMA RIAQFLSDIP ETVPLSTVNR QCSSGLQAVA SIAGGIRNGS YDIGMACGVE SMSLADRGNP GNITSRLMEK EKARDCLIPM GITSENVAER FGISREKQ Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 87%.

This gene encodes an enzyme operative in the beta-oxidation system of the peroxisomes. Deficiency of this enzyme leads to pseudo-Zellweger syndrome. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CL2663
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACAA1
Gene Accession No. P09110
Gene Alias 3-ketoacyl-CoA thiolase A, peroxisomal; 3-ketoacyl-CoA thiolase B, peroxisomal; 3-ketoacyl-CoA thiolase, peroxisomal; ACAA; ACAA1; Acaa1a; Acaa1b; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 1; acetyl-CoA acyltransferase 1A; acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A 1 peroxisomal; Acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A peroxisomal; acetyl-CoA acyltransferase A; Acetyl-CoA acyltransferase B; acetyl-CoA acyltransferase, 3-oxo acyl-CoA thiolase A; acetyl-Coenzyme A acyltransferase 1; acetyl-Coenzyme A acyltransferase 1 (peroxisomal 3-oxoacyl-Coenzyme A thiolase); acetyl-Coenzyme A acyltransferase 1A; acetyl-Coenzyme A acyltransferase 1B; beta-ketothiolase; Beta-ketothiolase A; beta-ketothiolase B; D9Ertd25e; peroxisomal 3-ketoacyl-CoA thiolase; peroxisomal 3-ketoacyl-CoA thiolase B; peroxisomal 3-oxoacyl-CoA thiolase; Peroxisomal 3-oxoacyl-CoA thiolase A; peroxisomal 3-oxoacyl-CoA thiolase B; peroxisomal 3-oxoacyl-Coenzyme A thiolase; Pktaa; PTHIO; PTL; RGD1562373; testicular tissue protein Li 197; THIO; thiolase A; thiolase B
Gene Symbols ACAA1
Host Species Mouse
Immunogen Recombinant protein fragment of Human ACAA1 (Product #RP-88949)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.