Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACAA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529957 Supplier Invitrogen™ Supplier No. PA529957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Chicken (83%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes an enzyme operative in the beta-oxidation system of the peroxisomes. Deficiency of this enzyme leads to pseudo-Zellweger syndrome. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ACAA1
Gene Accession No. P09110, Q8VCH0, Q921H8
Gene Alias 3-ketoacyl-CoA thiolase A, peroxisomal; 3-ketoacyl-CoA thiolase B, peroxisomal; 3-ketoacyl-CoA thiolase, peroxisomal; ACAA; ACAA1; Acaa1a; Acaa1b; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 1; acetyl-CoA acyltransferase 1A; acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A 1 peroxisomal; Acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A peroxisomal; acetyl-CoA acyltransferase A; Acetyl-CoA acyltransferase B; acetyl-CoA acyltransferase, 3-oxo acyl-CoA thiolase A; acetyl-Coenzyme A acyltransferase 1; acetyl-Coenzyme A acyltransferase 1 (peroxisomal 3-oxoacyl-Coenzyme A thiolase); acetyl-Coenzyme A acyltransferase 1A; acetyl-Coenzyme A acyltransferase 1B; beta-ketothiolase; Beta-ketothiolase A; beta-ketothiolase B; D9Ertd25e; peroxisomal 3-ketoacyl-CoA thiolase; peroxisomal 3-ketoacyl-CoA thiolase B; peroxisomal 3-oxoacyl-CoA thiolase; Peroxisomal 3-oxoacyl-CoA thiolase A; peroxisomal 3-oxoacyl-CoA thiolase B; peroxisomal 3-oxoacyl-Coenzyme A thiolase; Pktaa; PTHIO; PTL; RGD1562373; testicular tissue protein Li 197; THIO; thiolase A; thiolase B
Gene Symbols ACAA1, Acaa1a, Acaa1b
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ACAA1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113868, 235674, 30
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.