Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982347
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2347 Supplier Bioss Supplier No. BS12566R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an enzyme operative in the beta-oxidation system of the peroxisomes. Deficiency of this enzyme leads to pseudo-Zellweger syndrome. Alternative splicing results in multiple transcript variants.

Specifications

Antigen ACAA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ACAA1
Gene Accession No. P09110
Gene Alias 3-ketoacyl-CoA thiolase A, peroxisomal; 3-ketoacyl-CoA thiolase B, peroxisomal; 3-ketoacyl-CoA thiolase, peroxisomal; ACAA; Acaa1; Acaa1a; Acaa1b; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 1; acetyl-CoA acyltransferase 1A; acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A 1 peroxisomal; Acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A peroxisomal; Acetyl-CoA acyltransferase A; Acetyl-CoA acyltransferase B; acetyl-CoA acyltransferase, 3-oxo acyl-CoA thiolase A; acetyl-Coenzyme A acyltransferase 1; acetyl-Coenzyme A acyltransferase 1 (peroxisomal 3-oxoacyl-Coenzyme A thiolase); acetyl-Coenzyme A acyltransferase 1A; acetyl-Coenzyme A acyltransferase 1B; Beta-ketothiolase; Beta-ketothiolase A; Beta-ketothiolase B; D9Ertd25e; peroxisomal 3-ketoacyl-CoA thiolase; peroxisomal 3-ketoacyl-CoA thiolase B; Peroxisomal 3-oxoacyl-CoA thiolase; Peroxisomal 3-oxoacyl-CoA thiolase A; Peroxisomal 3-oxoacyl-CoA thiolase B; peroxisomal 3-oxoacyl-Coenzyme A thiolase; Pktaa; PTHIO; PTL; RGD1562373; testicular tissue protein Li 197; THIO; thiolase A; thiolase B
Gene Symbols ACAA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ABLIM3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.