Learn More
Invitrogen™ ACAA2 Monoclonal Antibody (OTI1B1)

Description
ACAA2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | ACAA2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1B1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ACAA2 |
| Gene Accession No. | P13437, P42765, Q8BWT1 |
| Gene Alias | 0610011L04Rik; 3-ketoacyl-CoA thiolase, mitochondrial; Acaa2; Acetyl-CoA acetyltransferase; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 2; acetyl-Coenzyme A acyltransferase 2; acetyl-Coenzyme A acyltransferase 2 (mitochondrial 3-oxoacyl-Coenzyme A thiolase); Acyl-CoA hydrolase, mitochondrial; AI255831; AI265397; beta ketothiolase; beta-ketothiolase; D18Ertd240e; DSAEC; mitochondrial 3-oxoacyl-CoA thiolase; mitochondrial 3-oxoacyl-Coenzyme A thiolase; T1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.