Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAA2 Monoclonal Antibody (OTI1B1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA506129

Description
3-Ketoacyl-CoA thiolase, mitochondrial also known as acetyl-Coenzyme A acyltransferase 2 is an enzyme that in humans is encoded by the ACAA2 gene. Acetyl-Coenzyme A acyltransferase 2 is an acetyl-CoA C-acyltransferase enzyme. The encoded protein catalyzes the last step of the mitochondrial fatty acid beta-oxidation spiral. Unlike most mitochondrial matrix proteins, it contains a non-cleavable amino-terminal targeting signal.Specifications
ACAA2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P13437, P42765, Q8BWT1 | |
ACAA2 | |
Full length human recombit protein of human ACAA2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B1 | |
Unconjugated | |
ACAA2 | |
DSAEC | |
Mouse | |
Affinity Chromatography | |
RUO | |
10449, 170465, 490568, 52538 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction