Learn More
ACAA2 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI2F6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506154

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
3-Ketoacyl-CoA thiolase, mitochondrial also known as acetyl-Coenzyme A acyltransferase 2 is an enzyme that in humans is encoded by the ACAA2 gene. Acetyl-Coenzyme A acyltransferase 2 is an acetyl-CoA C-acyltransferase enzyme. The encoded protein catalyzes the last step of the mitochondrial fatty acid beta-oxidation spiral. Unlike most mitochondrial matrix proteins, it contains a non-cleavable amino-terminal targeting signal.Specifications
ACAA2 | |
Monoclonal | |
Unconjugated | |
ACAA2 | |
DSAEC | |
Mouse | |
Affinity Chromatography | |
RUO | |
10449, 170465, 490568, 52538 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2F6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P13437, P42765, Q8BWT1 | |
ACAA2 | |
Full length human recombit protein of human ACAA2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.