Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAA2 Rabbit anti-Human, Mouse, Rat, Clone: 8H6H5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | ACAA2 |
---|---|
Clone | 8H6H5 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
ACAA2 Monoclonal antibody specifically detects ACAA2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
ACAA2 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10449 | |
IgG | |
Affinity purified |
8H6H5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
3-ketoacyl-CoA thiolase, mitochondrial, Acetyl-CoA acyltransferase, acetyl-CoA acyltransferase 2, acetyl-Coenzyme A acyltransferase 2, beta ketothiolase, beta-ketothiolase, DSAEC, EC 2.3.1, EC 2.3.1.16, FLJ35992, FLJ95265, Mitochondrial 3-oxoacyl-CoA thiolase, mitochondrial 3-oxoacyl-Coenzyme A thiolase, T1 | |
A synthetic peptide corresponding to a sequence within amino acids 298-397 of human ACAA2 (P42765). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title