Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACAA2 Recombinant Rabbit Monoclonal Antibody (12F4)
GREENER_CHOICE

Catalog No. FEMA550041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550041 Supplier Invitrogen™ Supplier No. MA550041
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

ACAA2 Recombinant Monoclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

ACAA2 (3-Ketoacyl-CoA thiolase) is an enzyme that in humans is encoded by the ACAA2 gene. Acetyl-Coenzyme A acyltransferase 2 is an acetyl-CoA C-acyltransferase enzyme. The encoded protein catalyzes the last step of the mitochondrial fatty acid beta-oxidation spiral. Unlike most mitochondrial matrix proteins, it contains a non-cleavable amino-terminal targeting signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 12F4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.4
Gene ACAA2
Gene Accession No. P42765, Q8BWT1
Gene Alias 0610011L04Rik; 3-ketoacyl-CoA thiolase, mitochondrial; Acaa2; Acetyl-CoA acetyltransferase; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 2; acetyl-Coenzyme A acyltransferase 2; acetyl-Coenzyme A acyltransferase 2 (mitochondrial 3-oxoacyl-Coenzyme A thiolase); Acyl-CoA hydrolase, mitochondrial; AI255831; AI265397; beta ketothiolase; beta-ketothiolase; D18Ertd240e; DSAEC; mitochondrial 3-oxoacyl-CoA thiolase; mitochondrial 3-oxoacyl-Coenzyme A thiolase; T1
Gene Symbols ACAA2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACAA2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10449, 52538
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.