Learn More
Invitrogen™ ACAA2 Recombinant Rabbit Monoclonal Antibody (8H6H5)

Description
Positive test controls include: HeLa, HepG2, A-549, Rat heart. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: PAISGALKKA GLSLKDMDLV EVNEAFAPQY LAVERSLDLD ISKTNVNGGA IALGHPLGGS GSRITAHLVH ELRRRGGKYA VGSACIGGGQ GIAVIIQSTA.
Specifications
Specifications
| Antigen | ACAA2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 8H6H5 |
| Concentration | 0.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | ACAA2 |
| Gene Accession No. | P13437, P42765, Q8BWT1 |
| Gene Alias | 0610011L04Rik; 3-ketoacyl-CoA thiolase, mitochondrial; Acaa2; Acetyl-CoA acetyltransferase; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 2; acetyl-Coenzyme A acyltransferase 2; acetyl-Coenzyme A acyltransferase 2 (mitochondrial 3-oxoacyl-Coenzyme A thiolase); Acyl-CoA hydrolase, mitochondrial; AI255831; AI265397; beta ketothiolase; beta-ketothiolase; D18Ertd240e; DSAEC; mitochondrial 3-oxoacyl-CoA thiolase; mitochondrial 3-oxoacyl-Coenzyme A thiolase; T1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.