Learn More
Invitrogen™ ACACB Polyclonal Antibody

Description
Immunogen sequence: ITKSKSEANL IPSQEPFPAS DNSGETPQRN GEGHTLPKTP SQAEPASHKG PKDAGRRRNS LPPSHQKPPR NPLSSSDAAP SPELQANGTG TQGLEATDTN GLSSSARPQG QQAGSPSKED KKQANIKRQL MTNFILGSFD DYSS Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 57%.
Specifications
Specifications
| Antigen | ACACB |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ACACB |
| Gene Accession No. | O00763 |
| Gene Alias | Acacb; Acc2; ACCB; ACC-beta; acetyl-CoA carboxylase 2; acetyl-CoA carboxylase beta; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta; AI597064; AW743042; Biotin carboxylase; HACC275 |
| Gene Symbols | ACACB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.