Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACACB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567926 Supplier Invitrogen™ Supplier No. PA567926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

Catalyzes the ATP-dependent carboxylation of acetyl-CoA to malonyl-CoA. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase. Involved in inhibition of fatty acid and glucose oxidation and enhancement of fat storage. May play a role in regulation of mitochondrial fatty acid oxidation through malonyl-CoA-dependent inhibition of carnitine palmitoyltransferase 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACACB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACACB
Gene Accession No. E9Q4Z2, O00763
Gene Alias Acacb; Acc2; ACCB; ACC-beta; acetyl-CoA carboxylase 2; acetyl-CoA carboxylase beta; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta; AI597064; AW743042; Biotin carboxylase; HACC275
Gene Symbols ACACB
Host Species Rabbit
Immunogen Synthesized peptide derived from human ACACB.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100705, 116719, 32
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.