Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACAD11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561711 Supplier Invitrogen™ Supplier No. PA561711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSLFYFWPRT VPMINQGSYS ENSGIPSMEE LISIYCRCRG INSILPNWNF FLALSYFKMA GIAQGVYSRY LLGNNSSEDS FLFANIVQPL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.

ACAD11 was isolated origainlly from rat liver peroxisomes. 34 known peroxisomal proteins have been identified in addition to several mitochondrial and microsomal proteins. ACAD11 is a bi-functional protein consisting of an aminoglycoside phosphotransferase-domain and an acyl-CoA dehydrogenase domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAD11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACAD11
Gene Accession No. Q709F0
Gene Alias 62113 protein; ACAD11; ACAD-11; Acyl-CoA dehydrogenase family member 11; acyl-CoA dehydrogenase family, member 11; acyl-Coenzyme A dehydrogenase family, member 11; FLJ12592; MGC150619
Gene Symbols ACAD11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACAD11. Recombinant protein control fragment (Product #RP-100574).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84129
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.