Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAD11 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$219.50 - $487.50
Specifications
Antigen | ACAD11 |
---|---|
Dilution | Western Blot |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ACAD11 Polyclonal antibody specifically detects ACAD11 in Human samples. It is validated for Western BlotSpecifications
ACAD11 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
ACAD-11, acyl-CoA dehydrogenase family member 11, acyl-CoA dehydrogenase family, member 11, acyl-Coenzyme A dehydrogenase family, member 11, EC 1.3.99.-, FLJ12592, MGC150619 | |
Synthetic peptide derived from the human ACAD11 protein (Uniprot: Q709F0) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
84129 | |
IgG | |
Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title