Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACAD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559250 Supplier Invitrogen™ Supplier No. PA559250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELFPVDVMRK AAQLGFGGVY IQTDVGGSGL SRLDTSVIFE ALATGCTSTT AYISIHNMCA WMIDSFGNEE QRHKFCPPL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

This gene encodes a member of the acyl-CoA dehydrogenase family of enzymes that catalyze the dehydrogenation of acyl-CoA derivatives in the metabolism of fatty acids or branch chained amino acids. The encoded protein is a mitochondrial enzyme that functions in catabolism of the branched-chain amino acid valine. Defects in this gene are the cause of isobutyryl-CoA dehydrogenase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAD8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACAD8
Gene Accession No. Q9UKU7
Gene Alias 2310016C19Rik; Acad8; ACAD-8; Activator-recruited cofactor 42 kDa component; acyl-CoA dehydrogenase family member 8; acyl-CoA dehydrogenase family, member 8; acyl-Coenzyme A dehydrogenase family, member 8; AI786953; ARC42; IBD; IBDH; Isobutyryl-CoA dehydrogenase, mitochondrial; RGD1564209
Gene Symbols ACAD8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACAD8. Recombinant protein control fragment (Product #RP-97418).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27034
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.