Learn More
Invitrogen™ ACAD9 Monoclonal Antibody (OTI1D4)

Description
ACAD9 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ACAD9 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1D4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Acad9 |
| Gene Accession No. | Q9H845 |
| Gene Alias | 2600017P15Rik; 4732402K02; ACAD9; ACAD-9; Acyl-CoA dehydrogenase family member 9; acyl-CoA dehydrogenase family member 9, mitochondrial; acyl-CoA dehydrogenase family, member 9; acyl-Coenzyme A dehydrogenase family, member 9; C630012L17Rik; Complex I assembly factor ACAD9, mitochondrial; NPD002; Nyggf2; very-long-chain acyl-CoA dehydrogenase VLCAD; very-long-chain acyl-CoA dehydrogenase VLCAD homolog |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.