Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACAD9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121139 Supplier Invitrogen™ Supplier No. PA5121139
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SKOV3, U-251MG, MCF7, 293T, NCI-H460, Mouse kidney, Mouse liver, Mouse brain. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: MSGCGLFLRT TAAARACRGL VVSTANRRLL RTSPPVRAFA KELFLGKIKK KEVFPFPEVS QDELNEINQF LGPVEKFFTE EVDSRKIDQE GKIPDETLEK LKSLGLFGLQ VPEEYGGLGF SNTMYSRLGE IISMDGSITV TLAAHQAIGL KGIILAGTEE QKAKYLPKLA SGEHIAAFCL TEPASGSDAA SIRSRATLSE DKKHYILNGS KVWITNGGLA NIFTVFAKTE VVDSDGSVKD KITAFIVERD FGGVTNGKPE DKLGIRGSNT.

This gene encodes a member of the acyl-CoA dehydrogenase family. Members of this family of proteins localize to the mitochondria and catalyze the rate-limiting step in the beta-oxidation of fatty acyl-CoA. The encoded protein is specifically active toward palmitoyl-CoA and long-chain unsaturated substrates. Mutations in this gene cause acyl-CoA dehydrogenase family member type 9 deficiency. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAD9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Acad9
Gene Accession No. Q8JZN5, Q9H845
Gene Alias 2600017P15Rik; 4732402K02; ACAD9; ACAD-9; Acyl-CoA dehydrogenase family member 9; acyl-CoA dehydrogenase family member 9, mitochondrial; acyl-CoA dehydrogenase family, member 9; acyl-Coenzyme A dehydrogenase family, member 9; C630012L17Rik; Complex I assembly factor ACAD9, mitochondrial; NPD002; Nyggf2; very-long-chain acyl-CoA dehydrogenase VLCAD; very-long-chain acyl-CoA dehydrogenase VLCAD homolog
Gene Symbols Acad9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ACAD9 (NP_054768.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229211, 28976
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.